Alfa Aesar™ Bis(benzonitrile)dichloroplatinum(II), Pt 40% min

Manufacturer: Alfa Aesar™ 1000406

Quantity 5g

 View more versions of this product

Catalog No. AA1000406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14873-63-3
    Formula Weight 472.25
    MDL Number MFCD00013125
    UN Number UN3439
    Chemical Name or Material Bis(benzonitrile)dichloroplatinum(II)

    Specifications

    Formula Weight 472.25
    Health Hazard 3 GHS P Statement
    P284a-P261-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00013125
    CAS 14873-63-3
    UN Number UN3439
    Physical Form Powder
    Health Hazard 1 Danger
    Quantity 5g
    Solubility Information Slightly soluble in acetone, chloroform
    Concentration Pt 40% min.
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H334-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material Bis(benzonitrile)dichloroplatinum(II)

    GHS H Statement
    H301-H311-H331-H334-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit