Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(benzonitrile)palladium(II) chloride, 97+%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
10g
Packaging:
Glass bottle
14220-64-5
C14H10Cl2N2Pd
383.55
MFCD00013123
WXNOJTUTEXAZLD-UHFFFAOYSA-L
bis benzonitrile palladium chloride,bis benzonitrile palladium ii chloride,trans-bis benzonitrile palladium ii chloride,bis benzonitrile palladium ii chloride, 500mg,palladium 2+ bis benzonitrile dichloride
10271322
benzonitrile;palladium(2+);dichloride
C1=CC=C(C=C1)C#N.C1=CC=C(C=C1)C#N.[Cl-].[Cl-].[Pd+2]
14220-64-5
C14H10Cl2N2Pd
383.55
MFCD00013123
WXNOJTUTEXAZLD-UHFFFAOYSA-L
bis benzonitrile palladium chloride,bis benzonitrile palladium ii chloride,trans-bis benzonitrile palladium ii chloride,bis benzonitrile palladium ii chloride, 500mg,palladium 2+ bis benzonitrile dichloride
10271322
benzonitrile;palladium(2+);dichloride
C1=CC=C(C=C1)C#N.C1=CC=C(C=C1)C#N.[Cl-].[Cl-].[Pd+2]
Elemental Analysis 99.0% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 120.0°C
Quantity 250mg
Color Orange to Brown
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 97+%
Linear Formula (C6H5CN)2PdCl2
Fieser 01,56; 05,31; 06,45; 09,44; 10,31; 13,34
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, toluene, chloroform
Formula Weight 383.55
Physical Form Crystalline Powder
Percent Purity 97+%
Chemical Name or Material Bis(benzonitrile)palladium(II) chloride
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 238-085-3