Learn More
Bis(carboxyethylgermanium) sesquioxide, 99.7%, Thermo Scientific Chemicals
Description
A bifunctional metalloligand with metal-C (Ge-C) bonds,bis(carboxyethylgermanium) sesquioxide, has been used to make two metal-organometallic frameworks (MOMFs). bis-(carboxyethylgermanium)sesquioxide is a as a promising anti-oxidative agent. Bis(carboxyethylgermanium) sesquioxide induce INF-γ and has undergone clinical trials with promising preclinical results.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 320°C (decomposition) |
Odor | Odorless |
Quantity | 5 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 339.25 |
Percent Purity | 99.7% |
Physical Form | Powder |
Chemical Name or Material | Bis(carboxyethylgermanium) sesquioxide |
Safety and Handling
EINECSNumber | 235-800-0 |
RTECSNumber | UA2459750 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.