Alfa Aesar™ Bis(cyclopentadienyl)dimethylhafnium(IV), 97+%

Manufacturer: Alfa Aesar™ H2696203

Quantity 1g

 View more versions of this product

Catalog No. AAH2696203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 37260-88-1
    Molecular Formula C12H16Hf
    Formula Weight 338.75
    MDL Number MFCD02093015
    Synonym Dimethylbis(cyclopentadienyl)hafnium(IV); Hafnocene dimethyl
    Chemical Name or Material Bis(cyclopentadienyl)dimethylhafnium(IV)

    Specifications

    Formula Weight 338.75
    Molecular Formula C12H16Hf
    MDL Number MFCD02093015
    CAS 37260-88-1
    Melting Point ≈118°C
    Synonym Dimethylbis(cyclopentadienyl)hafnium(IV); Hafnocene dimethyl
    Quantity 1g
    Chemical Name or Material Bis(cyclopentadienyl)dimethylhafnium(IV)
    Assay ≥97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit