Alfa Aesar™ Bis(cyclopentadienyl)magnesium, sublimed, 97%

Manufacturer: Alfa Aesar™ 4343403

Quantity 1g

 View more versions of this product

Catalog No. AA4343403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1284-72-6
    Molecular Formula C10H10Mg
    Formula Weight 154.49
    MDL Number MFCD00134487
    UN Number UN3391
    Chemical Name or Material Bis (cyclopentadienyl)magnesium, Sublimed

    Specifications

    Formula Weight 154.49
    Packaging Ampule
    Health Hazard 2 GHS H Statement
    H250-H252-H315-H319-H335
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P222-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 176°C
    Physical Form Powder
    Molecular Formula C10H10Mg
    CAS 1284-72-6
    Quantity 1g
    Health Hazard 1 Danger
    MDL Number MFCD00134487
    Sensitivity Air & Moisture Sensitive
    UN Number UN3391
    Color Off-white
    Boiling Point 290°C
    Storage Note 1
    • Ampouled under argon
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC
    Chemical Name or Material Bis (cyclopentadienyl)magnesium, Sublimed
    Assay 97%

    GHS H Statement
    H250-H252-H315-H319-H335
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P222-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit