Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bis(cyclopentadienyl)molybdenum dichloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
Description
Bis(cyclopentadienyl)molybdenum dichloride, 99% is used to prepare many derivatives. It is also used as pharmaceutical intermediates and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Quantity | 0.5 g |
UN Number | UN3261 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 297.04 |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | Bis(cyclopentadienyl)molybdenum dichloride |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
TSCA | No |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction