Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(cyclopentadienyl)nickel, dry

Manufacturer:  Alfa Aesar™ 8789909

Quantity 10g

 View more versions of this product

Catalog No. AA8789909

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1271-28-9
188.9
UN3391
Bis (cyclopentadienyl)nickel, Dry
C10H10Ni
MFCD00001441
Nickelocene; Di-pi-cyclopentadienylnickel

Specifications

1271-28-9
C10H10Ni
MFCD00001441
Dark green
Nickelocene; Di-pi-cyclopentadienylnickel
Air & Moisture Sensitive
Crystalline
188.9
UN3391
171° to 173°C
10g
Bis (cyclopentadienyl)nickel, Dry

Safety and Handling

SDS & Safety Documents

GHS H Statement
H250-H252-H301-H350-H317
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Toxic if swallowed.
May cause cancer.
May cause an allergic skin reaction.

GHS P Statement
P210-P222-P301+P310-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit