Alfa Aesar™ Bis(cyclopentadienyl)nickel, dry

Manufacturer: Alfa Aesar™ 8789918

Quantity 50g

 View more versions of this product

Catalog No. AA8789918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1271-28-9
    Molecular Formula C10H10Ni
    Formula Weight 188.9
    MDL Number MFCD00001441
    UN Number UN3391
    Synonym Nickelocene; Di-pi-cyclopentadienylnickel
    Chemical Name or Material Bis (cyclopentadienyl)nickel, Dry

    Specifications

    Formula Weight 188.9
    Health Hazard 3 GHS P Statement
    P210-P222-P301+P310-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not allow contact with air.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Physical Form Crystalline
    Color Dark green
    TSCA TSCA
    MDL Number MFCD00001441
    CAS 1271-28-9
    UN Number UN3391
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H250-H252-H301-H350-H317
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Toxic if swallowed.
    May cause cancer.
    May cause an allergic skin reaction.
    Melting Point 171° to 173°C
    Synonym Nickelocene; Di-pi-cyclopentadienylnickel
    Sensitivity Air & Moisture Sensitive
    Health Hazard 1 Danger
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC
    Molecular Formula C10H10Ni
    Chemical Name or Material Bis (cyclopentadienyl)nickel, Dry

    GHS H Statement
    H250-H252-H301-H350-H317
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Toxic if swallowed.
    May cause cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P222-P301+P310-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not allow contact with air.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit