Alfa Aesar™ Bis(cyclopentadienyl)titanium dichloride

Manufacturer: Alfa Aesar™ 1254722

Quantity 100g

 View more versions of this product

Catalog No. AA1254722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1271-19-8
    Formula Weight 249
    MDL Number MFCD00003723
    Merck Index 14,9482
    Synonym Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
    Chemical Name or Material Bis(cyclopentadienyl)titanium dichloride

    Specifications

    Formula Weight 249
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Storage Note 1
    • Packaged under argon
    RTECS Number XR2050000
    TSCA TSCA
    Physical Form Crystalline
    Merck Index 14,9482
    Quantity 100g
    Health Hazard 1 Warning
    Odor Odorless
    Density 1.60g/mL
    MDL Number MFCD00003723
    CAS 1271-19-8
    Synonym Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
    Sensitivity Air & Moisture Sensitive
    Color Red
    Melting Point ≈285°C (dec.)
    Health Hazard 2 GHS H Statement
    H341-H302-H332-H315-H319-H335
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Bis(cyclopentadienyl)titanium dichloride

    GHS H Statement
    H341-H302-H332-H315-H319-H335
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit