Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)titanium dichloride, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1271-19-8
C10H10Cl2Ti
249
MFCD00003723
MKNXBRLZBFVUPV-UHFFFAOYSA-L
Titanocene dichloride
124040768
cyclopenta-1,3-diene;titanium(2+);dichloride
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]
1271-19-8
C10H10Cl2Ti
249
MFCD00003723
MKNXBRLZBFVUPV-UHFFFAOYSA-L
Titanocene dichloride
124040768
cyclopenta-1,3-diene;titanium(2+);dichloride
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]
Specific Gravity 1.6
Color Bordeaux to Red
Melting Point 287.0°C to 289.0°C
Quantity 10g
Packaging Glass bottle
Density 1.6000g/mL
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (Argentometry)
Fieser 06,48
Merck Index 15, 9638
Solubility Information Solubility in water: hydrolyses. Other solubilities: soluble in aromatic hydrocarbons, thf, halogenated, solvents, acetonitrile,, slightly soluble in chloroform, ethanol < 1mg/l,, acetone < 1mg/l
Formula Weight 249
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Bis(cyclopentadienyl)titanium dichloride, 97%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Cal

GHS Signal Word: Warning

EINECSNumber : 215-035-9

RTECSNumber : XR2050000

TSCA : TSCA