Alfa Aesar™ Bis(cyclopentadienyl)tungsten dihydride, 98%

Manufacturer: Alfa Aesar™ 4426603

Quantity 1g

 View more versions of this product

Catalog No. AA4426603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1271-33-6
    Molecular Formula C10H12W
    Formula Weight 316.06
    MDL Number MFCD04126433
    UN Number UN3396
    Chemical Name or Material Bis (cyclopentadienyl)tungsten dihydride

    Specifications

    Formula Weight 316.06
    Packaging Ampule
    Color Yellow
    Melting Point >350°C
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Molecular Formula C10H12W
    MDL Number MFCD04126433
    CAS 1271-33-6
    UN Number UN3396
    Quantity 1g
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE
    Physical Form Crystalline
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H228-H261-H315-H319-H335
    Flammable solid.
    In contact with water releases flammable gas.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Bis (cyclopentadienyl)tungsten dihydride
    Assay 98%

    GHS H Statement
    H228-H261-H315-H319-H335
    Flammable solid.
    In contact with water releases flammable gas.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P231+P232-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    Avoid breathing dust/fume/gas/mist/vapors/spray.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit