Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)vanadium, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1277-47-0
C5H6V+2
117.044
MFCD00058735
JRYGAVLSRSKLBL-UHFFFAOYSA-N
bis cyclopentadienyl vanadium
131698989
cyclopenta-1,3-diene;vanadium(2+)
C1C=CC=C1.[V+2]
1277-47-0
C5H6V+2
117.044
MFCD00058735
JRYGAVLSRSKLBL-UHFFFAOYSA-N
bis cyclopentadienyl vanadium
131698989
cyclopenta-1,3-diene;vanadium(2+)
C1C=CC=C1.[V+2]
Quantity 1g
Packaging Glass bottle
Color Black
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Merck Index 15, 10111
Formula Weight 181.13
Physical Form Crystalline Powder
Percent Purity ≥94% (permanganometry)
Chemical Name or Material Bis(cyclopentadienyl)vanadium
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic in contact with skin.
Toxic if inhaled.
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger