Alfa Aesar™ Bis(cyclopentadienyl)vanadium dichloride, 95%

Manufacturer: Alfa Aesar™ 3958806

Quantity 5g

 View more versions of this product

Catalog No. AA3958806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12083-48-6
    Formula Weight 252.04
    MDL Number MFCD00014260
    UN Number UN3285
    Synonym Vanadocene dichloride
    Chemical Name or Material Bis(cyclopentadienyl)vanadium dichloride

    Specifications

    Formula Weight 252.04
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    UN Number UN3285
    Sensitivity Air & Moisture Sensitive
    Health Hazard 1 Danger
    Odor Pungent
    RTECS Number YW1580000
    Physical Form Crystalline
    Melting Point 250°C (dec.)
    Synonym Vanadocene dichloride
    Quantity 5g
    Density 1.60g/mL
    MDL Number MFCD00014260
    CAS 12083-48-6
    Color Green
    Storage Note 1
    • Packaged under argon
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
    Chemical Name or Material Bis(cyclopentadienyl)vanadium dichloride
    Assay 95%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit