Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Bis(cyclopentadienyl)vanadium dichloride, 95%
Click to view available options
Quantity:
1g
Description
- Packaged under argon
Specifications
Density | 1.60 g/mL |
Sensitivity | Air and moisture sensitive |
Odor | Pungent |
Melting Point | 250°C (decomposition) |
Color | Green |
Quantity | 1g |
UN Number | UN3285 |
Solubility Information | Reacts with water |
Formula Weight | 252.04 |
Physical Form | Crystalline |
Show More |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
EINECSNumber : 235-150-8
RTECSNumber : YW1580000
TSCA : No
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen