Alfa Aesar™ Bis(cyclopentadienyl)vanadium, sublimed, 95%

Manufacturer: Alfa Aesar™ 39572MD

Quantity 250mg

 View more versions of this product

Catalog No. AA39572MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1277-47-0
    Formula Weight 181.13
    MDL Number MFCD00058735
    UN Number UN3181
    Synonym Vanadocene
    Chemical Name or Material Bis(cyclopentadienyl)vanadium, Sublimed

    Specifications

    Formula Weight 181.13
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    UN Number UN3181
    Sensitivity Air & Moisture Sensitive
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H228-H315-H319-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00058735
    CAS 1277-47-0
    Boiling Point 200°C (1mmHg) (subl.)
    Synonym Vanadocene
    Health Hazard 1 Danger
    Melting Point 165° to 167°C
    Storage Note 1
    • Packaged under argon
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    Quantity 250mg
    Chemical Name or Material Bis(cyclopentadienyl)vanadium, Sublimed
    Assay 95%

    GHS H Statement
    H300-H310-H330-H228-H315-H319-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit