Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)zirconium chloride hydride, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
37342-97-5
C10H11ClZr
257.86
MFCD02089401
VPQMECJLNPUNGN-UHFFFAOYSA-M
zirconocene chloride hydride
131718033
cyclopenta-1,3-diene;hydride;zirconium(2+);chloride
[H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2]
37342-97-5
C10H11ClZr
257.86
MFCD02089401
VPQMECJLNPUNGN-UHFFFAOYSA-M
zirconocene chloride hydride
131718033
cyclopenta-1,3-diene;hydride;zirconium(2+);chloride
[H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2]
Reduction reagent
Packaging Glass bottle
Quantity 5g
Melting Point >300.0°C
Infrared Spectrum Authentic
Color White to Beige
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (Argentometry)
Merck Index 15, 8533
Solubility Information Solubility in water: reacts with water.
Formula Weight 257.86
Physical Form Powder
Percent Purity 95%
Chemical Name or Material Bis(cyclopentadienyl)zirconium chloride hydride
Show More Show Less

GHS H Statement
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g

GHS Signal Word: Danger

EINECSNumber : 253-479-5