Alfa Aesar™ Bis(cyclopentadienyl)zirconium dichloride

Manufacturer: Alfa Aesar™ 1254822

Quantity 100g

 View more versions of this product

Catalog No. AA1254822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1291-32-3
    Formula Weight 292.32
    MDL Number MFCD00003726
    Synonym Zirconocene dichloride
    Chemical Name or Material Bis(cyclopentadienyl)zirconium dichloride

    Specifications

    Formula Weight 292.32
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number ZH7525000
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00003726
    Melting Point 242° to 245°C
    Boiling Point 124° to 125°C (15mmHg)
    CAS 1291-32-3
    Color White to off-white
    Synonym Zirconocene dichloride
    Sensitivity Air & Moisture Sensitive
    Health Hazard 1 Warning
    Quantity 100g
    Storage Note 1
    • Packaged under argon
    Chemical Name or Material Bis(cyclopentadienyl)zirconium dichloride

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit