Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)zirconium dichloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1291-32-3
C10H10Cl2Zr
292.32
MFCD00003726
Zirconocene dichloride
1291-32-3
C10H10Cl2Zr
292.32
MFCD00003726
Zirconocene dichloride
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Melting Point 242.0°C to 245.0°C
Color White to Beige
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (Argentometry)
Fieser 11,51; 16,393
Solubility Information Solubility in water: hydrolysis.
Formula Weight 292.32
Physical Form Crystalline Powder or Crystals
Percent Purity 98+%
Chemical Name or Material Bis(cyclopentadienyl)zirconium dichloride
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 215-066-8