Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(dicarbonylcyclopentadienyliron), 99%

Manufacturer:  Alfa Aesar™ 3111414

Quantity 25g

 View more versions of this product

Catalog No. AA3111414


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12154-95-9
MFCD00135625
Dicarbonylcyclopentadienyliron dimer; Dicyclopentadienyl-di-iron tetracarbonyl
353.93
UN3466
Bis(dicarbonylcyclopentadienyliron)

Specifications

12154-95-9
353.93
UN3466
Red
25g
Bis(dicarbonylcyclopentadienyliron)
Crystalline
MFCD00135625
Dicarbonylcyclopentadienyliron dimer; Dicyclopentadienyl-di-iron tetracarbonyl
194°C (dec.)
Air & Moisture Sensitive
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H300-H310-H330
Flammable solid.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.

storageNote1 :

  • Packaged under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit