Alfa Aesar™ Bis(dicarbonylcyclopentadienyliron), 99%

Manufacturer: Alfa Aesar™ 3111422

Quantity 100g

 View more versions of this product

Catalog No. AA3111422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12154-95-9
    Formula Weight 353.93
    MDL Number MFCD00135625
    UN Number UN3466
    Synonym Dicarbonylcyclopentadienyliron dimer; Dicyclopentadienyl-di-iron tetracarbonyl
    Chemical Name or Material Bis(dicarbonylcyclopentadienyliron)

    Specifications

    Formula Weight 353.93
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00135625
    Melting Point 194°C (dec.)
    Health Hazard 2 GHS H Statement
    H228-H300-H310-H330
    Flammable solid.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Physical Form Crystalline
    CAS 12154-95-9
    Color Red
    Health Hazard 1 Danger
    Quantity 100g
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    UN Number UN3466
    Synonym Dicarbonylcyclopentadienyliron dimer; Dicyclopentadienyl-di-iron tetracarbonyl
    Sensitivity Air & Moisture Sensitive
    Storage Note 1
    • Packaged under argon
    Chemical Name or Material Bis(dicarbonylcyclopentadienyliron)
    Assay 99%

    GHS H Statement
    H228-H300-H310-H330
    Flammable solid.
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.

    GHS P Statement
    P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit