Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(dichlorophosphino)methane, 96%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
28240-68-8
CH2Cl4P2
217.79
MFCD01073767
OEKLCAXLANZKPX-UHFFFAOYSA-N
bis dichlorophosphino methane,dichloro dichlorophosphanyl methyl phosphane,acmc-1cq46,methylenebis dichlorophosphine,methylenebis phosphonous dichloride,dichloro dichlorophosphanylmethyl phosphane,phosphonous dichloride,p,p'-methylenebis
2784185
dichloro(dichlorophosphanylmethyl)phosphane
C(P(Cl)Cl)P(Cl)Cl
28240-68-8
CH2Cl4P2
217.79
MFCD01073767
OEKLCAXLANZKPX-UHFFFAOYSA-N
bis dichlorophosphino methane,dichloro dichlorophosphanyl methyl phosphane,acmc-1cq46,methylenebis dichlorophosphine,methylenebis phosphonous dichloride,dichloro dichlorophosphanylmethyl phosphane,phosphonous dichloride,p,p'-methylenebis
2784185
dichloro(dichlorophosphanylmethyl)phosphane
C(P(Cl)Cl)P(Cl)Cl
NMR Complies, 95% min.
Flash Point 96°C
Boiling Point 101.0°C to 105.0°C (22.0 mmHg)
Density 1.6010g/mL
Quantity 1g
Packaging Glass bottle
Refractive Index 1.5920 to 1.5950
Specific Gravity 1.601
Color Colorless to Yellow
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (Argentometry)
Linear Formula (Cl2P)2CH2
Solubility Information Solubility in water: reacts
Formula Weight 217.79
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Bis(dichlorophosphino)methane
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger