Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(dichlorophosphino)methane, 96%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
28240-68-8
CH2Cl4P2
217.79
MFCD01073767
OEKLCAXLANZKPX-UHFFFAOYSA-N
bis dichlorophosphino methane,dichloro dichlorophosphanyl methyl phosphane,acmc-1cq46,methylenebis dichlorophosphine,methylenebis phosphonous dichloride,dichloro dichlorophosphanylmethyl phosphane,phosphonous dichloride,p,p'-methylenebis
2784185
dichloro(dichlorophosphanylmethyl)phosphane
C(P(Cl)Cl)P(Cl)Cl
28240-68-8
CH2Cl4P2
217.79
MFCD01073767
OEKLCAXLANZKPX-UHFFFAOYSA-N
bis dichlorophosphino methane,dichloro dichlorophosphanyl methyl phosphane,acmc-1cq46,methylenebis dichlorophosphine,methylenebis phosphonous dichloride,dichloro dichlorophosphanylmethyl phosphane,phosphonous dichloride,p,p'-methylenebis
2784185
dichloro(dichlorophosphanylmethyl)phosphane
C(P(Cl)Cl)P(Cl)Cl
Boiling Point 101.0°C to 105.0°C (22.0 mmHg)
Packaging Glass bottle
NMR Complies, 95% min.
Refractive Index 1.5920 to 1.5950
Flash Point 96°C
Specific Gravity 1.601
Quantity 5g
Density 1.6010g/mL
Color Colorless to Yellow
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula (Cl2P)2CH2
Solubility Information Solubility in water: reacts
Formula Weight 217.79
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Bis(dichlorophosphino)methane
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger