Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(diethylamino)phenylphosphine, Acros Organics™

Click to view available options
Quantity:
5g
25g
1636-14-2
C14H25N2P
252.34
HTIVDCMRYKVNJC-UHFFFAOYSA-N
phenylbis diethylamino phosphine,bis diethylamino phenylphosphine,diethylamino phenyl phosphanyl diethylamine,acmc-20almr,tetraethylphenylphosphonous diamide,phenylphosphonous tetraethyldiamide,bis diethylamino phenylphosphine 97,n-diethylamino phenyl phosphanyl-n-ethylethanamine,phosphonous diamide,n,n,n',n'-tetraethyl-p-phenyl
137129
N-[diethylamino(phenyl)phosphanyl]-N-ethylethanamine
CCN(CC)P(C1=CC=CC=C1)N(CC)CC
1636-14-2
C14H25N2P
252.34
HTIVDCMRYKVNJC-UHFFFAOYSA-N
phenylbis diethylamino phosphine,bis diethylamino phenylphosphine,diethylamino phenyl phosphanyl diethylamine,acmc-20almr,tetraethylphenylphosphonous diamide,phenylphosphonous tetraethyldiamide,bis diethylamino phenylphosphine 97,n-diethylamino phenyl phosphanyl-n-ethylethanamine,phosphonous diamide,n,n,n',n'-tetraethyl-p-phenyl
137129
N-[diethylamino(phenyl)phosphanyl]-N-ethylethanamine
CCN(CC)P(C1=CC=CC=C1)N(CC)CC
Density 0.9710g/mL
Specific Gravity 0.971
Flash Point 106°C
Quantity 5g
Linear Formula C6H5P(N(CH2CH3)2)2
Formula Weight 252.34
Chemical Name or Material Bis(diethylamino)phenylphosphine