Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(diethylamino)phenylphosphine, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
1636-14-2
C14H25N2P
252.34
HTIVDCMRYKVNJC-UHFFFAOYSA-N
phenylbis diethylamino phosphine, bis diethylamino phenylphosphine, diethylamino phenyl phosphanyl diethylamine, acmc-20almr, tetraethylphenylphosphonous diamide, phenylphosphonous tetraethyldiamide, bis diethylamino phenylphosphine 97, n-diethylamino phenyl phosphanyl-n-ethylethanamine, phosphonous diamide,n,n,n',n'-tetraethyl-p-phenyl
137129
N-[diethylamino(phenyl)phosphanyl]-N-ethylethanamine
CCN(CC)P(C1=CC=CC=C1)N(CC)CC
1636-14-2
C14H25N2P
252.34
HTIVDCMRYKVNJC-UHFFFAOYSA-N
phenylbis diethylamino phosphine, bis diethylamino phenylphosphine, diethylamino phenyl phosphanyl diethylamine, acmc-20almr, tetraethylphenylphosphonous diamide, phenylphosphonous tetraethyldiamide, bis diethylamino phenylphosphine 97, n-diethylamino phenyl phosphanyl-n-ethylethanamine, phosphonous diamide,n,n,n',n'-tetraethyl-p-phenyl
137129
N-[diethylamino(phenyl)phosphanyl]-N-ethylethanamine
CCN(CC)P(C1=CC=CC=C1)N(CC)CC

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Density 0.9710g/mL
Flash Point 106°C
Linear Formula C6H5P(N(CH2CH3)2)2
Quantity 25g
Specific Gravity 0.971
Formula Weight 252.34
Chemical Name or Material Bis(diethylamino)phenylphosphine

RUO – Research Use Only