Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(diethylamino)phosphorochloridate, 97%

Click to view available options
Quantity:
5g
25g
1794-24-7
C8H20ClN2OP
226.685
MFCD00137958
HKKBJRUSFKTHSU-UHFFFAOYSA-N
bis diethylamino phosphoryl chloride,tetraethylphosphorodiamidic chloride,bis diethylamino chlorophosphine oxide,didiethylaminophosphinoyl chloride,n,n,n',n'-tetraethylphosphorodiamidic chloride,phosphorodiamidic chloride, tetraethyl,tetraethylphosphorodiamidic acid chloride,bis diethylamino phosphochloridate,bis diethylamino phosphinoyl chloride,phosphorodiamidicchloride, n,n,n',n'-tetraethyl
74528
N-[chloro(diethylamino)phosphoryl]-N-ethylethanamine
CCN(CC)P(=O)(N(CC)CC)Cl
1794-24-7
C8H20ClN2OP
226.685
MFCD00137958
HKKBJRUSFKTHSU-UHFFFAOYSA-N
bis diethylamino phosphoryl chloride,tetraethylphosphorodiamidic chloride,bis diethylamino chlorophosphine oxide,didiethylaminophosphinoyl chloride,n,n,n',n'-tetraethylphosphorodiamidic chloride,phosphorodiamidic chloride, tetraethyl,tetraethylphosphorodiamidic acid chloride,bis diethylamino phosphochloridate,bis diethylamino phosphinoyl chloride,phosphorodiamidicchloride, n,n,n',n'-tetraethyl
74528
N-[chloro(diethylamino)phosphoryl]-N-ethylethanamine
CCN(CC)P(=O)(N(CC)CC)Cl
Boiling Point 101°C (0.3mmHg)
Quantity 5g
UN Number UN3265
Formula Weight 226.69
Percent Purity 97%
Chemical Name or Material Bis(diethylamino)phosphorochloridate

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.