Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(dimethylamino)phosphorochloridate, 90%, Tech., ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1605-65-8
C4H12ClN2OP
170.58
MFCD00008302
WYLQARGYFXBZMD-UHFFFAOYSA-N
bis dimethylamino phosphoryl chloride,phosphorodiamidic chloride, tetramethyl,bis dimethylamino phosphinic chloride,n,n,n',n'-tetramethylphosphorodiamidic chloride,bis dimethylamino phosphorylchloride,bis n,n-dimethylamino phosphinic chloride,ch3 2n 2pocl,bis dimethylamino phosphorochloridate,tetramethylphosphorodiamidic chloride,phosphorodiamidicchloride, n,n,n',n'-tetramethyl
74150
N-[chloro(dimethylamino)phosphoryl]-N-methylmethanamine
CN(C)P(=O)(N(C)C)Cl
1605-65-8
C4H12ClN2OP
170.58
MFCD00008302
WYLQARGYFXBZMD-UHFFFAOYSA-N
bis dimethylamino phosphoryl chloride,phosphorodiamidic chloride, tetramethyl,bis dimethylamino phosphinic chloride,n,n,n',n'-tetramethylphosphorodiamidic chloride,bis dimethylamino phosphorylchloride,bis n,n-dimethylamino phosphinic chloride,ch3 2n 2pocl,bis dimethylamino phosphorochloridate,tetramethylphosphorodiamidic chloride,phosphorodiamidicchloride, n,n,n',n'-tetramethyl
74150
N-[chloro(dimethylamino)phosphoryl]-N-methylmethanamine
CN(C)P(=O)(N(C)C)Cl
Boiling Point 246.0°C to 247.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Density 1.1500g/mL
Flash Point 119°C
Refractive Index 1.4650 to 1.4670
Specific Gravity 1.15
Quantity 10g
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88 to 112% (ex Cl) (Argentometry)
Linear Formula [(CH3)2N]2P(O)Cl
Formula Weight 170.58
Physical Form Liquid
Percent Purity 90%
Grade Technical
Chemical Name or Material Bis(dimethylamino)phosphorochloridate
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

EINECSNumber : 216-517-1