Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(dimethylamino)phosphorochloridate, 94%

Manufacturer:  Alfa Aesar™ 3927409

Quantity 10g

 View more versions of this product

Catalog No. AA3927409

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1605-65-8
170.58
UN3265
Bis(dimethylamino)phosphoryl chloride; N,N,N'N'-Tetramethylphosphorodiamidic chloride
[(CH3)2N]2P(O)Cl
MFCD00008302
471430
Bis(dimethylamino)phosphorochloridate

Specifications

1605-65-8
1.4679
170.58
UN3265
Bis(dimethylamino)phosphoryl chloride; N,N,N'N'-Tetramethylphosphorodiamidic chloride
Colorless
10g
Pungent
Bis(dimethylamino)phosphorochloridate
Liquid
[(CH3)2N]2P(O)Cl
MFCD00008302
471430
1.152g/mL
246° to 247°C
119° (246°F)
Moisture Sensitive
94%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

storageNote1 :

  • Packaged under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit