Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(diphenylphosphino)acetylene, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
5112-95-8
C26H20P2
394.394
MFCD00003045
FOWZHJNBFVLJGP-UHFFFAOYSA-N
bis diphenylphosphino acetylene,phosphine, ethynylenebis diphenyl,ethynylenebis diphenylphosphine,phosphine, 1,2-ethynediylbis diphenyl,1,2-bis diphenylphosphino ethyne,2-diphenylphosphanylethynyl diphenyl phosphane,2-diphenylphosphanyl ethynyl diphenylphosphane,ethyne-1,2-diylbis diphenylphosphane,acmc-20almm,pubchem6917
78789
2-diphenylphosphanylethynyl(diphenyl)phosphane
C1=CC=C(C=C1)P(C#CP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
5112-95-8
C26H20P2
394.394
MFCD00003045
FOWZHJNBFVLJGP-UHFFFAOYSA-N
bis diphenylphosphino acetylene,phosphine, ethynylenebis diphenyl,ethynylenebis diphenylphosphine,phosphine, 1,2-ethynediylbis diphenyl,1,2-bis diphenylphosphino ethyne,2-diphenylphosphanylethynyl diphenyl phosphane,2-diphenylphosphanyl ethynyl diphenylphosphane,ethyne-1,2-diylbis diphenylphosphane,acmc-20almm,pubchem6917
78789
2-diphenylphosphanylethynyl(diphenyl)phosphane
C1=CC=C(C=C1)P(C#CP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
Quantity 1g
Color White
Packaging Glass bottle
Melting Point 84.0°C to 88.0°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C6H5)2PC≡CP(C6H5)2
Formula Weight 394.39
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material Bis(diphenylphosphino)acetylene, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 225-842-8