Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(ethylcyclopentadienyl)magnesium, 98+%

Manufacturer:  Alfa Aesar™ 3859706

Quantity 5g

 View more versions of this product

Catalog No. AA3859706


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

114460-02-5
MFCD00151375
Bis(ethylcyclopentadienyl)magnesium
210.6
UN3392

Specifications

114460-02-5
Liquid
MFCD00151375
1.91g/mL
Air & Moisture Sensitive
≥98%
Ampule
210.6
UN3392
5g
Bis(ethylcyclopentadienyl)magnesium

Safety and Handling

SDS & Safety Documents

GHS H Statement
H250-H252-H315-H319-H335
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit