Alfa Aesar™ Bis(ethylcyclopentadienyl)magnesium, 98+%

Manufacturer: Alfa Aesar™ 3859703

Quantity 1g

 View more versions of this product

Catalog No. AA3859703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 114460-02-5
    Formula Weight 210.6
    MDL Number MFCD00151375
    UN Number UN3392
    Chemical Name or Material Bis(ethylcyclopentadienyl)magnesium

    Specifications

    Formula Weight 210.6
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P210-P222-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Health Hazard 2 GHS H Statement
    H250-H252-H315-H319-H335
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00151375
    UN Number UN3392
    Density 1.91g/mL
    Health Hazard 1 Danger
    Quantity 1g
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC
    Physical Form Liquid
    CAS 114460-02-5
    Chemical Name or Material Bis(ethylcyclopentadienyl)magnesium
    Assay ≥98%

    GHS H Statement
    H250-H252-H315-H319-H335
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P222-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit