Alfa Aesar™ Bis(hydroxymethyl)phenylphosphine

Manufacturer: Alfa Aesar™ H27733ME

Quantity 500mg

 View more versions of this product

Catalog No. AAH27733ME

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3127-08-0
    Molecular Formula C6H5P(CH2OH)2
    Formula Weight 170.15
    MDL Number MFCD01311823
    UN Number UN2811
    Synonym (Phenylphosphinidene)dimethanol
    Chemical Name or Material Bis(hydroxymethyl)phenylphosphine

    Specifications

    Formula Weight 170.15
    Molecular Formula C6H5P(CH2OH)2
    MDL Number MFCD01311823
    CAS 3127-08-0
    UN Number UN2811
    Synonym (Phenylphosphinidene)dimethanol
    Melting Point 56° to 58°C
    Health Hazard 1 Danger
    Quantity 500mg
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material Bis(hydroxymethyl)phenylphosphine

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit