Alfa Aesar™ Bis(isopropylcyclopentadienyl)tungsten dihydride, 98+%

MOCVD precursor

Manufacturer: Alfa Aesar™ 4267204

Quantity 2g

 View more versions of this product

Catalog No. AA4267204

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64561-25-7
    Formula Weight 400.22
    MDL Number MFCD04126431
    UN Number UN3398
    Chemical Name or Material Bis (isopropylcyclopentadienyl)tungsten dihydride

    Specifications

    Formula Weight 400.22
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 64561-25-7
    UN Number UN3398
    Health Hazard 1 Warning
    Quantity 2g
    MDL Number MFCD04126431
    Boiling Point 124°C (0.1mmHg)
    Sensitivity Air & Moisture Sensitive
    Storage Note 1
    • Ampouled under argon
    DOT Information Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE
    Physical Form Liquid
    Color Orange
    Health Hazard 2 GHS H Statement
    H226-H261-H315-H319-H335
    Flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Bis (isopropylcyclopentadienyl)tungsten dihydride
    Assay ≥98%

    GHS H Statement
    H226-H261-H315-H319-H335
    Flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit