Alfa Aesar™ Bis(mercaptocyclohexane)titanium tetrachloride, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 3954006

Quantity 5g

 View more versions of this product

Catalog No. AA3954006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 149391-23-1
    Formula Weight 422.1
    MDL Number MFCD00171702
    UN Number UN3261
    Synonym Tetrachlorobis(cyclohexylmercapto)titanium(IV)
    Chemical Name or Material Bis(mercaptocyclohexane)titanium tetrachloride

    Specifications

    Formula Weight 422.1
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.
    UN Number UN3261
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    MDL Number MFCD00171702
    CAS 149391-23-1
    Melting Point 76° to 80°C
    Health Hazard 1 Danger
    Physical Form Crystalline
    Synonym Tetrachlorobis(cyclohexylmercapto)titanium(IV)
    Color Yellow
    Sensitivity Moisture Sensitive
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Quantity 5g
    Chemical Name or Material Bis(mercaptocyclohexane)titanium tetrachloride
    Assay 99.9%, (metals basis)

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit