Alfa Aesar™ Bis(methylcyclopentadienyl)nickel(II), 97%

Manufacturer: Alfa Aesar™ H2746403

Quantity 1g

 View more versions of this product

Catalog No. AAH2746403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1293-95-4
    Molecular Formula C12H14Ni
    Formula Weight 216.93
    MDL Number MFCD01863654
    Synonym 1,1'-Dimethylnickelocene
    Chemical Name or Material Bis(methylcyclopentadienyl)nickel(II)

    Specifications

    Formula Weight 216.93
    Molecular Formula C12H14Ni
    MDL Number MFCD01863654
    CAS 1293-95-4
    Synonym 1,1'-Dimethylnickelocene
    Melting Point 34° to 36°C
    Boiling Point 85° to 90°C (1mmHg)
    Quantity 1g
    Flash Point 66°C (151°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H351-H332-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Suspected of causing cancer.
    Harmful if inhaled.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Chemical Name or Material Bis(methylcyclopentadienyl)nickel(II)
    Assay 97%

    GHS H Statement
    H301-H311-H351-H332-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Suspected of causing cancer.
    Harmful if inhaled.
    May cause an allergic skin reaction.

    2° to 8°C

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit