Alfa Aesar™ Bis(methylcyclopentadienyl)zirconium dichloride, 98%

Manufacturer: Alfa Aesar™ H2724103

Quantity 1g

 View more versions of this product

Catalog No. AAH2724103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12109-71-6
    Molecular Formula C12H14Cl2Zr
    Formula Weight 320.37
    MDL Number MFCD00672106
    UN Number UN3261
    Synonym 1,1'-Dimethylzirconocene dichloride
    Chemical Name or Material Bis(methylcyclopentadienyl)zirconium dichloride

    Specifications

    Formula Weight 320.37
    TSCA TSCA
    Molecular Formula C12H14Cl2Zr
    MDL Number MFCD00672106
    UN Number UN3261
    Synonym 1,1'-Dimethylzirconocene dichloride
    Melting Point 180° to 184°C
    Quantity 1g
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    CAS 12109-71-6
    Health Hazard 2 GHS H Statement
    H260-H314-H318
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material Bis(methylcyclopentadienyl)zirconium dichloride
    Assay 98%

    GHS H Statement
    H260-H314-H318
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit