Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(methylcyclopentadienyl)zirconium dichloride, 98%

Manufacturer:  Alfa Aesar™ H2724106

Quantity 5g

 View more versions of this product

Catalog No. AAH2724106


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12109-71-6
320.37
UN3261
Bis(methylcyclopentadienyl)zirconium dichloride
C12H14Cl2Zr
MFCD00672106
1,1'-Dimethylzirconocene dichloride

Specifications

12109-71-6
320.37
UN3261
180° to 184°C
Bis(methylcyclopentadienyl)zirconium dichloride
C12H14Cl2Zr
MFCD00672106
1,1'-Dimethylzirconocene dichloride
5g
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H260-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit