Alfa Aesar™ Bis(n-propyltetramethylcyclopentadienyl)barium

Manufacturer: Alfa Aesar™ 3857402

Quantity 0.5g

 View more versions of this product

Catalog No. AA3857402

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 210758-43-3
    Formula Weight 463.9
    MDL Number MFCD00151385
    UN Number UN1564
    Chemical Name or Material Bis(n-propyltetramethylcyclopentadienyl)barium

    Specifications

    Physical Form Viscous Liquid/Solid
    Formula Weight 463.9
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P261-P304+P340-P311-P301+P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00151385
    CAS 210758-43-3
    Health Hazard 2 GHS H Statement
    H331-H302
    Toxic if inhaled.
    Harmful if swallowed.
    UN Number UN1564
    Quantity 0.5g
    Health Hazard 1 Danger
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Chemical Name or Material Bis(n-propyltetramethylcyclopentadienyl)barium

    GHS H Statement
    H331-H302
    Toxic if inhaled.
    Harmful if swallowed.

    GHS P Statement
    P261-P304+P340-P311-P301+P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit