Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(norbornadiene)rhodium(I) tetrafluoroborate, 94%, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
Packaging:
Glass bottle
36620-11-8
C14H16BF4Rh
373.99
MFCD00671775
ZWPWNBFOFVFMPP-UHFFFAOYSA-N
bis norbornadiene rhodium i tetrafluoroborate,rh nbd 2bf4,bis eta-2,5-norbornadiene rhodium i tetrafluoroborate,bis norbornadiene tetrafluoroborato rhodium,bis bicyclo 2.2.1 hepta-2,5-diene rhodium tetrafluoroborate,rh nbd 2 bf4,bis norbomadiene rhodium i tetrafluoroborate,bis norbornadiene rhodium tetrafluoroborate
10915722
bicyclo[2.2.1]hepta-2,5-diene;rhodium;tetrafluoroborate
[B-](F)(F)(F)F.C1C2C=CC1C=C2.C1C2C=CC1C=C2.[Rh]
36620-11-8
C14H16BF4Rh
373.99
MFCD00671775
ZWPWNBFOFVFMPP-UHFFFAOYSA-N
bis norbornadiene rhodium i tetrafluoroborate,rh nbd 2bf4,bis eta-2,5-norbornadiene rhodium i tetrafluoroborate,bis norbornadiene tetrafluoroborato rhodium,bis bicyclo 2.2.1 hepta-2,5-diene rhodium tetrafluoroborate,rh nbd 2 bf4,bis norbomadiene rhodium i tetrafluoroborate,bis norbornadiene rhodium tetrafluoroborate
10915722
bicyclo[2.2.1]hepta-2,5-diene;rhodium;tetrafluoroborate
[B-](F)(F)(F)F.C1C2C=CC1C=C2.C1C2C=CC1C=C2.[Rh]
Color Red
Melting Point 209.0°C to 211.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500mg
Elemental Analysis 92% min.
Assay Percent Range 94%
Solubility Information Solubility in water: decomposes
Formula Weight 373.99
Physical Form Powder
Percent Purity 94%
Chemical Name or Material Bis(norbornadiene)rhodium(I) tetrafluoroborate
Show More Show Less

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning