Alfa Aesar™ Bis(p-toluenesulfonyl)methane, 97%

Manufacturer: Alfa Aesar™ H26617MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH26617MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15310-28-8
    Molecular Formula C15H16O4S2
    Formula Weight 324.42
    MDL Number MFCD00032255
    Synonym Methylene bis(p-tolyl sulfone)
    Chemical Name or Material Bis(p-toluenesulfonyl)methane

    Specifications

    Formula Weight 324.42
    Synonym Methylene bis(p-tolyl sulfone)
    Melting Point 137° to 139°C
    Quantity 250mg
    Molecular Formula C15H16O4S2
    MDL Number MFCD00032255
    CAS 15310-28-8
    Chemical Name or Material Bis(p-toluenesulfonyl)methane
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit