Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(pentafluorophenyl) carbonate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
59483-84-0
C13F10O3
394.124
MFCD00368353
IOVVFSGCNWQFQT-UHFFFAOYSA-N
bis pentafluorophenyl carbonate,bis perfluorophenyl carbonate,dipentafluorophenylcarbonate,pentafluorophenyl carbonate,di-pentafluorophenyl carbonate,bis 2,3,4,5,6-pentafluorophenyl carbonate,carbonic acid bis pentafluorophenyl ester,dpfpc,acmc-209mds
2734833
bis(2,3,4,5,6-pentafluorophenyl) carbonate
C1(=C(C(=C(C(=C1F)F)F)F)F)OC(=O)OC2=C(C(=C(C(=C2F)F)F)F)F
59483-84-0
C13F10O3
394.124
MFCD00368353
IOVVFSGCNWQFQT-UHFFFAOYSA-N
bis pentafluorophenyl carbonate,bis perfluorophenyl carbonate,dipentafluorophenylcarbonate,pentafluorophenyl carbonate,di-pentafluorophenyl carbonate,bis 2,3,4,5,6-pentafluorophenyl carbonate,carbonic acid bis pentafluorophenyl ester,dpfpc,acmc-209mds
2734833
bis(2,3,4,5,6-pentafluorophenyl) carbonate
C1(=C(C(=C(C(=C1F)F)F)F)F)OC(=O)OC2=C(C(=C(C(=C2F)F)F)F)F
Coupling reagent
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Melting Point 48°C to 51°C
Color White
Assay Percent Range 98%
Formula Weight 394.12
Physical Form Crystalline Powder and Chunks
Percent Purity ≥97.5%
Chemical Name or Material Bis(pentafluorophenyl) carbonate