Alfa Aesar™ Bis(pentamethylcyclopentadienyl)barium 1,2-dimethoxyethane adduct

Manufacturer: Alfa Aesar™ 4363903

Quantity 1g

 View more versions of this product

Catalog No. AA4363903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 312739-90-5
    Molecular Formula C20H30Ba·C4H10O2
    Formula Weight 479.92
    MDL Number MFCD02684523
    UN Number UN1564
    Chemical Name or Material Bis (pentamethylcyclopentadienyl)barium 1,2-dimethoxyethane adduct

    Specifications

    Formula Weight 479.92
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C20H30Ba·C4H10O2
    CAS 312739-90-5
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Physical Form Powder
    MDL Number MFCD02684523
    UN Number UN1564
    Quantity 1g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Health Hazard 1 Warning
    Storage Note 1
    • Ampouled under argon
    Chemical Name or Material Bis (pentamethylcyclopentadienyl)barium 1,2-dimethoxyethane adduct

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit