Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(pentamethylcyclopentadienyl)chromium, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
74507-61-2
C20 H30 Cr
MFCD00058886
74507-61-2
C20 H30 Cr
MFCD00058886
Quantity 1g
Color Black
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 322.45
Physical Form Crystalline Powder
Chemical Name or Material Bis(pentamethylcyclopentadienyl)chromium

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Recommended Storage : Freezer -20°C