Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(pentamethylcyclopentadienyl)iron, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
12126-50-0
C20H30Fe
MFCD00058736
Decamethylferrocene
12126-50-0
C20H30Fe
MFCD00058736
Decamethylferrocene
Color Yellow to Orange
Packaging Glass bottle
Quantity 5g
NMR Authentic
Formula Weight 326.3
Physical Form Crystalline Powder
Chemical Name or Material Bis(pentamethylcyclopentadienyl)iron