Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Bis(pentamethylcyclopentadienyl)manganese, 98%
Click to view available options
Quantity:
1g
5g
Specifications
Quantity | 5g |
Sensitivity | Air and moisture sensitive |
Melting Point | 230°C (decomposition) |
Packaging | Ampule |
Formula Weight | 325.38 |
Physical Form | Crystalline |
Percent Purity | 98% |
Chemical Name or Material | Bis(pentamethylcyclopentadienyl)manganese |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures