Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(pentamethylcyclopentadienyl)titanium dichloride, 99%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
11136-36-0
C20H30Cl2Ti
389.24
MFCD00049153
ZBFBXTFQCKIUHU-UHFFFAOYSA-L
titanium 4+ chloride 2,3,4,5,5-pentamethylcyclopenta-1,3-dien-1-ide 1/2/2
91997586
1,2,3,5,5-pentamethylcyclopenta-1,3-diene;titanium(4+);dichloride
CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Cl-].[Cl-].[Ti+4]
11136-36-0
C20H30Cl2Ti
389.24
MFCD00049153
ZBFBXTFQCKIUHU-UHFFFAOYSA-L
titanium 4+ chloride 2,3,4,5,5-pentamethylcyclopenta-1,3-dien-1-ide 1/2/2
91997586
1,2,3,5,5-pentamethylcyclopenta-1,3-diene;titanium(4+);dichloride
CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Cl-].[Cl-].[Ti+4]
Melting Point 190°C
Quantity 1g
Color Brown or Red
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98.5% min. (Argentometry)
Solubility Information Solubility in water: reacts
Formula Weight 389.24
Physical Form Crystals or Needles
Percent Purity 99%
Chemical Name or Material Bis(pentamethylcyclopentadienyl)titanium dichloride
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning