Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(pentamethylcyclopentadienyl)zirconium dichloride, 99%

Manufacturer:  Alfa Aesar™ 3955502

Quantity 0.5g

 View more versions of this product

Catalog No. AA3955502

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

54039-38-2
MFCD00058849
Decamethylzirconocene dichloride
432.56
UN3096
Bis(pentamethylcyclopentadienyl)zirconium dichloride

Specifications

54039-38-2
Crystalline
MFCD00058849
Decamethylzirconocene dichloride
0.5g
Bis(pentamethylcyclopentadienyl)zirconium dichloride
Ampule
432.56
UN3096
Pale yellow
Air & Moisture Sensitive
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H261-H301-H311-H314-H318-H332
In contact with water releases flammable gas.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement
P231+P232-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Handle under inert gas.
Protect from moisture.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, WATER-REACTIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit