Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(phenylsulfonyl)methane, 99%, Thermo Scientific™

Catalog Number p-69196
Click to view available options
Quantity:
1 g
5 g
3406-02-8
C13H12O4S2
296.37
MFCD00007553
QCHNSJNRFSOCLJ-UHFFFAOYSA-N
2-phenylsulfonyl acetophenone, 2-phenylsulfonylacetophenone, 1-phenyl-2-phenylsulfonyl ethanone, acetophenone, 2-phenylsulfonyl, 2-benzenesulfonyl-1-phenylethanone, ethanone, 1-phenyl-2-phenylsulfonyl, 1-phenyl-2-phenylsulphonyl ethan-1-one, alpha-phenylsulfonylacetophenone, 2-benzenesulfonyl-1-phenylethan-1-one, 1-phenyl-2-phenylsulfonyl ethan-1-one
76949
(benzenesulfonyl)methanesulfonylbenzene
O=S(=O)(CS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1
3406-02-8
C13H12O4S2
296.37
MFCD00007553
QCHNSJNRFSOCLJ-UHFFFAOYSA-N
2-phenylsulfonyl acetophenone, 2-phenylsulfonylacetophenone, 1-phenyl-2-phenylsulfonyl ethanone, acetophenone, 2-phenylsulfonyl, 2-benzenesulfonyl-1-phenylethanone, ethanone, 1-phenyl-2-phenylsulfonyl, 1-phenyl-2-phenylsulphonyl ethan-1-one, alpha-phenylsulfonylacetophenone, 2-benzenesulfonyl-1-phenylethan-1-one, 1-phenyl-2-phenylsulfonyl ethan-1-one
76949
(benzenesulfonyl)methanesulfonylbenzene
O=S(=O)(CS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1
Melting Point 121°C to 122°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula (C6H5SO2)2CH2
Quantity 1 g
Formula Weight 296.37
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Bis(phenylsulfonyl)methane, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 222-291-5

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit