Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(pyridine)iodonium tetrafluoroborate, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
15656-28-7
C10H10IN2·BF4
371.91
MFCD03703393
WVXJCTSZJWIBQT-UHFFFAOYSA-N
bis pyridine iodonium tetrafluoroborate,barluenga's reagent,bis pyridine iodonium i tetrafluoroborate,iodanium pyridine tetrafluoroborate,iodonium pyridine tetrafluoroborate,bispyridine iodonium tetrafluoroborate,bis pyridin iodonium tetrafluoroborate,iodium bis pyridine tetrafluoroborate,bis pyridine-iodonium tetrafluoroborate
10883201
iodanium;pyridine;tetrafluoroborate
[B-](F)(F)(F)F.C1=CC=NC=C1.C1=CC=NC=C1.[IH2+]
15656-28-7
C10H10IN2·BF4
371.91
MFCD03703393
WVXJCTSZJWIBQT-UHFFFAOYSA-N
bis pyridine iodonium tetrafluoroborate,barluenga's reagent,bis pyridine iodonium i tetrafluoroborate,iodanium pyridine tetrafluoroborate,iodonium pyridine tetrafluoroborate,bispyridine iodonium tetrafluoroborate,bis pyridin iodonium tetrafluoroborate,iodium bis pyridine tetrafluoroborate,bis pyridine-iodonium tetrafluoroborate
10883201
iodanium;pyridine;tetrafluoroborate
[B-](F)(F)(F)F.C1=CC=NC=C1.C1=CC=NC=C1.[IH2+]
C-X bond formation reagent
Packaging Glass bottle
Melting Point 137.0°C to 141.0°C
Quantity 1g
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Merck Index 15, 1303
Formula Weight 371.91
Physical Form Powder
Percent Purity 97%
Chemical Name or Material Bis(pyridine)iodonium tetrafluoroborate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning