Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Bis[(R)-1-phenylethyl]amine, ChiPros™, 99%, ee 98+%, Thermo Scientific Chemicals

Catalog Number aah2742403
Click to view available options
Quantity:
1 g
5 g
25 g
23294-41-9
C16H19N
225.34
MFCD00243088
NXLACVVNHYIYJN-UHFFFAOYNA-N
+-bis r-1-phenylethyl amine, r-bis r-1-phenylethyl amine, bis r-alpha-methylbenzyl amine, +-bis r-1-phenylethy amine hydrochloride, 1r-1-phenyl-n-1r-1-phenylethyl ethanamine, bis 1r-1-phenylethyl amine, r,r-bis-1-phenylethyl amin, +-bis-r-1-phenylethyl amine hydrochloride
5702599
(1R)-1-phenyl-N-[(1R)-1-phenylethyl]ethanamine
CC(NC(C)C1=CC=CC=C1)C1=CC=CC=C1
23294-41-9
C16H19N
225.34
MFCD00243088
NXLACVVNHYIYJN-UHFFFAOYNA-N
+-bis r-1-phenylethyl amine, r-bis r-1-phenylethyl amine, bis r-alpha-methylbenzyl amine, +-bis r-1-phenylethy amine hydrochloride, 1r-1-phenyl-n-1r-1-phenylethyl ethanamine, bis 1r-1-phenylethyl amine, r,r-bis-1-phenylethyl amin, +-bis-r-1-phenylethyl amine hydrochloride
5702599
(1R)-1-phenyl-N-[(1R)-1-phenylethyl]ethanamine
CC(NC(C)C1=CC=CC=C1)C1=CC=CC=C1

(+)-Bis[(R)-1-phenylethyl]amine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-Bis[(R)-1-phenylethyl]amine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 98°C (0.3 mmHg)
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 225.34
Percent Purity 99%
Assay ee ≥98%
Chemical Name or Material (+)-Bis[(R)-1-phenylethyl]amine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit