Alfa Aesar™ Bis(tetramethylcyclopentadienyl)chromium(II), 98+%

Manufacturer: Alfa Aesar™ H2739603

Quantity 1g

 View more versions of this product

Catalog No. AAH2739603

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 82066-37-3
    Molecular Formula C18H26Cr
    Formula Weight 294.41
    MDL Number MFCD01862451
    Chemical Name or Material Bis(tetramethylcyclopentadienyl)chromium(II)

    Specifications

    Formula Weight 294.41
    Molecular Formula C18H26Cr
    MDL Number MFCD01862451
    CAS 82066-37-3
    Melting Point 106° to 111°C
    Quantity 1g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Chemical Name or Material Bis(tetramethylcyclopentadienyl)chromium(II)
    Assay ≥98%

    GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.

    2° to 8°C

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit